Learn More
TARGETMOL CHEMICALS INC ABEMACICLIB METHANESULFO 100MG
SDP
Catalog No. 502576997
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Abemaciclib methanesulfonate (LY2835219) is a specific and effective inhibitor of CDK4(IC50=2 nM) and CDK6(IC50=10 nM). purity: 99%
Catalog No. 50-257-6997 Supplier TARGETMOL CHEMICALS INC Supplier No. T3111100MG
Also available in 2 mg 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Abemaciclib methanesulfonate (LY2835219) is a specific and effective inhibitor of CDK4(IC50=2 nM) and CDK6(IC50=10 nM). purity: 99%