Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abemaciclib (methanesulfonate) | 1231930-82-7 | 99.8% | 602.70 | 1ML
SDP

Catalog No. 5000391914
Encompass_Preferred

Abemaciclib methanesulfonate is a selective and BBB-permeable CDK4/6 inhibitor. It exhibits IC50s of 2 nM for CDK4 and 10 nM for CDK6. This product is suitable for use as a laboratory chemical and in the manufacture of substances.

  • Selective and BBB-permeable CDK4/6 inhibitor
  • IC50s of 2 nM for CDK4 and 10 nM for CDK6
  • Suitable for laboratory chemical applications
  • Can be used in the manufacture of substances
  • Appears as a solid
  • Recommended storage at 4°C, sealed and away from moisture
  • Solvent storage at -80°C for 2 years or -20°C for 1 year
  • Ships at room temperature if less than 2 weeks

Catalog No. 50-003-91914 Supplier Medchemexpress LLC Supplier No. HY1629710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Abemaciclib methanesulfonate is a selective and BBB-permeable CDK4/6 inhibitor. It exhibits IC50s of 2 nM for CDK4 and 10 nM for CDK6. This product is suitable for use as a laboratory chemical and in the manufacture of substances.

  • Selective and BBB-permeable CDK4/6 inhibitor
  • IC50s of 2 nM for CDK4 and 10 nM for CDK6
  • Suitable for laboratory chemical applications
  • Can be used in the manufacture of substances
  • Appears as a solid
  • Recommended storage at 4°C, sealed and away from moisture
  • Solvent storage at -80°C for 2 years or -20°C for 1 year
  • Ships at room temperature if less than 2 weeks

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.