Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abemaciclib methanesulfonate (LY2835219 methanesulfonate) | 1231930-82-7 | 99.8% | 50 MG
SDP

Catalog No. 5000366010
Encompass_Preferred

Abemaciclib methanesulfonate (LY2835219 methanesulfonate) | 1231930-82-7 | 99.8% | 50 MG

Catalog No. 50-003-66010 Supplier Medchemexpress LLC Supplier No. 50MGHY16297
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Abemaciclib methanesulfonate (LY2835219 methanesulfonate) | 1231930-82-7 | 99.8% | 50 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.