Learn More
Medchemexpress LLC Abeprazan hydrochloride | 1902954-87-3 | 99.9% | 446.87 | 25 MG
SDP
Catalog No. 5000347513
Encompass_Preferred
Abeprazan hydrochloride (DWP14012 hydrochloride) is a potassium-competitive acid blocker that inhibits H+, K+- ATPase by reversible potassium-competitive ionic binding with no acid activation required. It is developed as a potential alternative to proton pump inhibitors for treating acid-related diseases.
  • Potassium-competitive acid blocker
  • Inhibits H+, K+- ATPase by reversible potassium-competitive ionic binding
  • No acid activation required for inhibition
  • Potential alternative to proton pump inhibitors for acid-related diseases
Catalog No. 50-003-47513 Supplier Medchemexpress LLC Supplier No. 25MGHY109079A
Abeprazan hydrochloride (DWP14012 hydrochloride) is a potassium-competitive acid blocker that inhibits H+, K+- ATPase by reversible potassium-competitive ionic binding with no acid activation required. It is developed as a potential alternative to proton pump inhibitors for treating acid-related diseases.
  • Potassium-competitive acid blocker
  • Inhibits H+, K+- ATPase by reversible potassium-competitive ionic binding
  • No acid activation required for inhibition
  • Potential alternative to proton pump inhibitors for acid-related diseases