Learn More
Medchemexpress LLC Abeprazan hydrochloride | 1902954-87-3 | 99.9% | 446.87 | 50 MG
SDP
Catalog No. 5000344408
Encompass_Preferred
Abeprazan hydrochloride (DWP14012 hydrochloride) is a potassium-competitive acid blocker that inhibits H+, K+- ATPase through reversible potassium-competitive ionic binding without requiring acid activation. It is being developed as a potential alternative to proton pump inhibitors for treating acid-related diseases.
  • Potassium-competitive acid blocker.
  • Inhibits H+, K+- ATPase by reversible potassium-competitive ionic binding.
  • Does not require acid activation for its inhibitory mechanism.
  • Developed as a potential alternative for proton pump inhibitors.
  • Inhibits acid secretion in a dose-dependent manner in in vivo studies.
  • Efficacy is equal to or greater than that of vonoprazan in various in vivo studies.
Catalog No. 50-003-44408 Supplier Medchemexpress LLC Supplier No. HY109079A10MM/1ML
Abeprazan hydrochloride (DWP14012 hydrochloride) is a potassium-competitive acid blocker that inhibits H+, K+- ATPase through reversible potassium-competitive ionic binding without requiring acid activation. It is being developed as a potential alternative to proton pump inhibitors for treating acid-related diseases.
  • Potassium-competitive acid blocker.
  • Inhibits H+, K+- ATPase by reversible potassium-competitive ionic binding.
  • Does not require acid activation for its inhibitory mechanism.
  • Developed as a potential alternative for proton pump inhibitors.
  • Inhibits acid secretion in a dose-dependent manner in in vivo studies.
  • Efficacy is equal to or greater than that of vonoprazan in various in vivo studies.