Learn More
TARGETMOL CHEMICALS INC ABETA-IN-1 5MG
SDP
Catalog No. 502579433
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. A(beta)-IN-1 is a novel potent and orally active (gamma)-secretase modulator (GSM). A(beta)-IN-1 potently reduced A(beta)42 levels with an IC 50 value of 0.091 uM in cultured cells without inhibiting CYP3A4. A(beta)-IN-1 shows a sustained pharmacokinetic profile. purity: 98%
Catalog No. 50-257-9433 Supplier TARGETMOL CHEMICALS INC Supplier No. T632085MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. A(beta)-IN-1 is a novel potent and orally active (gamma)-secretase modulator (GSM). A(beta)-IN-1 potently reduced A(beta)42 levels with an IC 50 value of 0.091 uM in cultured cells without inhibiting CYP3A4. A(beta)-IN-1 shows a sustained pharmacokinetic profile. purity: 98%