Learn More
Enzo Life Sciences Amyloid beta Recombinant monoclonal antibody (6E10) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6120200
Encompass_Preferred
Clone: 6E10. Isotype: IgGκ. Immunogen: Amyloid-beta peptide. Species reactivity: Human. Specificity: Recognizes aa 1-16 of β-amyloid. Applications: ELISA, IHC, IP, WB Electron microscopy. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3138