Learn More
Bachem Amyloid Beta Protein (42-1)
SDP
Supplier:  Bachem 4027991.0500
Encompass_Preferred
Amyloid Beta Protein (42-1), H-Ala-Ile-Val-Val-Gly-Gly-Val-Met-Leu-Gly-Ile-Ile-Ala-Gly-Lys-Asn-Ser-Gly-Val-Asp-Glu-Ala-Phe-Phe-Val-Leu-Lys-Gln-His-His-Val-Glu-Tyr-Gly-Ser-Asp-His-Arg-Phe-Glu-Ala-Asp-OH, 317366-82-8, C203H311N55O60S, Mr 4514.10, 0.5mg. b-Amyloid (42-1). Reverse sequence of Ab 1-42, inactive control for the Ab.
Catalog No. NC1418235