Learn More
TARGETMOL CHEMICALS INC ABETA42-IN-2 5MG
SDP
Catalog No. 502578989
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A(beta)42-IN-2 is a (gamma)-secretase modulator. A(beta)42-IN-2 has an IC 50 of 6.5 nM for (alpha)(beta) 42. A(beta)42-IN-2 can be used for the Alzheimer's disease research[1]. purity: 99%
Catalog No. 50-257-8989 Supplier TARGETMOL CHEMICALS INC Supplier No. T96415MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. A(beta)42-IN-2 is a (gamma)-secretase modulator. A(beta)42-IN-2 has an IC 50 of 6.5 nM for (alpha)(beta) 42. A(beta)42-IN-2 can be used for the Alzheimer's disease research[1]. purity: 99%