Learn More
Medchemexpress LLC HY-10990 5mg Medchemexpress, Abexinostat CAS:783355-60-2 Purity:>98%
SDP
Catalog No. 501968726
Encompass_Preferred
Medchemexpress, HY-10990 5mg Abexinostat CAS:783355-60-2 Abexinostat (CRA 024781) is a novel pan-HDAC inhibitor mostly targeting HDAC1 with Ki of 7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8726 Supplier Medchemexpress LLC Supplier No. HY109905MG
Medchemexpress, HY-10990 5mg Abexinostat CAS:783355-60-2 Abexinostat (CRA 024781) is a novel pan-HDAC inhibitor mostly targeting HDAC1 with Ki of 7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.