Learn More
Selleck Chemical LLC Abexinostat (PCI-24781) S1090-5mg
SDP
Catalog No. 503108141
Encompass_Preferred
Abexinostat (PCI-24781 CRA-024781) is a novel pan-HDAC inhibitor mostly targeting HDAC1 with Ki of 7 nM modest potent to HDACs 2 3 6 and 10 and greater than 40-fold selectivity against HDAC8 Phase 1/2
Catalog No. 50-310-8141 Supplier Selleck Chemical LLC Supplier No. S10905MG
Abexinostat (PCI-24781 CRA-024781) is a novel pan-HDAC inhibitor mostly targeting HDAC1 with Ki of 7 nM modest potent to HDACs 2 3 6 and 10 and greater than 40-fold selectivity against HDAC8 Phase 1/2