Learn More
ABclonal Technology ABflo 488 anti-Mouse CD80 mAb
SDP
Supplier:  ABclonal Technology A26801200T
Encompass_Preferred
Predicted to enable coreceptor activity Acts upstream of or within T cell costimulation cellular response to lipopolysaccharide and positive regulation of alpha-beta T cell proliferation Located in external side of plasma membrane Is expressed in heart ventricle and skin Human ortholog(s) of this gene implicated in type 1 diabetes mellitus Orthologous to human CD80 (CD80 molecule)
Catalog No. 50-000-08866