Learn More
ABclonal Technology ABflo 488 Rabbit anti- CD10
SDP
Catalog No. 5000012470
Encompass_Preferred
Enables peptidase activity Involved in amyloid-beta clearance positive regulation of long-term synaptic potentiation and sensory perception of pain Acts upstream of or within amyloid-beta metabolic process Located in several cellular components including dendrite neuron projection terminus and synaptic vesicle Is expressed in several structures including alimentary system central nervous system genitourinary system liver and biliary system and mammary gland Used to study Alzheimer s disease Human ortholog(s) of this gene implicated in Alzheimer s disease Charcot-Marie-Tooth disease axonal type 2T cerebellar ataxia type 43 cerebral amyloid angiopathy and membranous glomerulonephritis Orthologous to human MME (membrane metalloendopeptidase)
Catalog No. 50-000-12470 Supplier ABclonal Technology Supplier No. A25704500T
Enables peptidase activity Involved in amyloid-beta clearance positive regulation of long-term synaptic potentiation and sensory perception of pain Acts upstream of or within amyloid-beta metabolic process Located in several cellular components including dendrite neuron projection terminus and synaptic vesicle Is expressed in several structures including alimentary system central nervous system genitourinary system liver and biliary system and mammary gland Used to study Alzheimer s disease Human ortholog(s) of this gene implicated in Alzheimer s disease Charcot-Marie-Tooth disease axonal type 2T cerebellar ataxia type 43 cerebral amyloid angiopathy and membranous glomerulonephritis Orthologous to human MME (membrane metalloendopeptidase)