Learn More
ABclonal Technology ABflo 488 Rabbit anti- IL-13
SDP
Catalog No. 5000010359
Encompass_Preferred
Predicted to enable interleukin-13 receptor binding activity Involved in positive regulation of cold-induced thermogenesis and positive regulation of macrophage activation Acts upstream of or within cellular response to cytokine stimulus inflammatory response and positive regulation of immune effector process Located in cytoplasm external side of plasma membrane and extracellular space Is expressed in gut liver male reproductive gland or organ and thymus Used to study atopic dermatitis Human ortholog(s) of this gene implicated in several diseases including allergic disease (multiple) autoimmune disease (multiple) diffuse scleroderma lung disease (multiple) and nose disease (multiple) Orthologous to human IL13 (interleukin 13)
Catalog No. 50-000-10359 Supplier ABclonal Technology Supplier No. A25118100T
Predicted to enable interleukin-13 receptor binding activity Involved in positive regulation of cold-induced thermogenesis and positive regulation of macrophage activation Acts upstream of or within cellular response to cytokine stimulus inflammatory response and positive regulation of immune effector process Located in cytoplasm external side of plasma membrane and extracellular space Is expressed in gut liver male reproductive gland or organ and thymus Used to study atopic dermatitis Human ortholog(s) of this gene implicated in several diseases including allergic disease (multiple) autoimmune disease (multiple) diffuse scleroderma lung disease (multiple) and nose disease (multiple) Orthologous to human IL13 (interleukin 13)