Learn More
ABclonal Technology ABflo 488 Rat anti-Mouse CD122
SDP
Catalog No. 5000014137
Encompass_Preferred
The interleukin 2 receptor is composed of alpha and beta subunits The beta subunit encoded by this gene is very homologous to the human beta subunit and also shows structural similarity to other cytokine receptors
Catalog No. 50-000-14137 Supplier ABclonal Technology Supplier No. A27775100UG
The interleukin 2 receptor is composed of alpha and beta subunits The beta subunit encoded by this gene is very homologous to the human beta subunit and also shows structural similarity to other cytokine receptors