Learn More
ABclonal Technology ABflo 594 Rabbit anti-Rat CD31
SDP
Supplier:  ABclonal Technology A25154100T
Encompass_Preferred
Enables protein phosphatase binding activity Involved in several processes including cellular response to interferon-gamma negative regulation of GTPase activity and negative regulation of actin filament polymerization Located in cell surface and ruffle Used to study pulmonary hypertension Biomarker of bacterial pneumonia orchitis and osteoarthritis Human ortholog(s) of this gene implicated in coronary artery disease (multiple) lung non-small cell carcinoma neuroblastoma and psoriatic arthritis Orthologous to human PECAM1 (platelet and endothelial cell adhesion molecule 1)
Catalog No. 50-000-10438