Learn More
ABclonal Technology ABflo 647 Rabbit anti- CD59a
SDP
Catalog No. 5000007831
Encompass_Preferred
Predicted to enable complement binding activity Involved in several processes including negative regulation of angiogenesis negative regulation of fibroblast growth factor production and negative regulation of vascular endothelial growth factor receptor signaling pathway Acts upstream of or within negative regulation of activation of membrane attack complex Located in external side of plasma membrane Is expressed in several structures including Meckel s cartilage nervous system neural retina skeleton and testis Human ortholog(s) of this gene implicated in colorectal carcinoma and ovarian cancer Orthologous to human CD59 (CD59 molecule (CD59 blood group))
Catalog No. 50-000-07831 Supplier ABclonal Technology Supplier No. A23362500T
Predicted to enable complement binding activity Involved in several processes including negative regulation of angiogenesis negative regulation of fibroblast growth factor production and negative regulation of vascular endothelial growth factor receptor signaling pathway Acts upstream of or within negative regulation of activation of membrane attack complex Located in external side of plasma membrane Is expressed in several structures including Meckel s cartilage nervous system neural retina skeleton and testis Human ortholog(s) of this gene implicated in colorectal carcinoma and ovarian cancer Orthologous to human CD59 (CD59 molecule (CD59 blood group))