Learn More
ABclonal Technology ABflo700 Rat anti-Mouse CD3
SDP
Catalog No. 5000009702
Encompass_Preferred
This gene encodes a component of the alpha-beta T cell receptor complex involved in T cell activation and thymic T cell selection It is predicted to have transmembrane signaling receptor activity and protein-binding functions It plays roles in cytokine production signal transduction nervous system development and cell adhesion The protein is located at the plasma membrane dendritic spines and immunological synapses It is expressed in the thymus colon and hemolymphoid system Human orthologs CD3D and CD3E are associated with immunodeficiencies including SCID and immunodeficiencies 18 and 19
Catalog No. 50-000-09702 Supplier ABclonal Technology Supplier No. A27432200T
This gene encodes a component of the alpha-beta T cell receptor complex involved in T cell activation and thymic T cell selection It is predicted to have transmembrane signaling receptor activity and protein-binding functions It plays roles in cytokine production signal transduction nervous system development and cell adhesion The protein is located at the plasma membrane dendritic spines and immunological synapses It is expressed in the thymus colon and hemolymphoid system Human orthologs CD3D and CD3E are associated with immunodeficiencies including SCID and immunodeficiencies 18 and 19