Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ VersiPlate Frame, 96-well, skirted, white

Catalog No. AB1805 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1805 25 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1805 Supplier Thermo Scientific™ Supplier No. AB1805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flexible 96-well plates of low profile strip tubes for use in PCR and qPCR applications.

The Thermo Scientific VersiPlates are unique, with each strip of eight tubes being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.

Highlights
  • Easier sample handling as multiple strips of samples can be handled simultaneously
  • Reduced consumables waste as only the required numbers of strips need to be removed with the remainder being stored for the next experiment
  • Can be used in instruments with or without a VersiFrame
  • The reusable polycarbonate frame can serve as a useful rack for a VersiPlate or any number of low profile strip tubes and facilitates sample loading into thermal cyclers
  • The low profile format of the VersiPlate has a lower dead volume which reduces sample evaporation
  • Available in white which improves signal reflection, reduces variation and increases the sensitivity of qPCR assays
  • Can be used with all Thermo Scientific VersiCap™ Mats, cap strips, adhesive and heat seals
Applications
  • PCR
  • qPCR/Real-Time PCR
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Packaging 25 Plate
Product Line VersiPlate
Color White
Product Type Frame
For Use With (Equipment) Real-Time PCR Systems or Thermal Cyclers
Material Polycarbonate
No. of Wells 96
Plate Compatibility 96-Well Plates
Quantity 25 plates
Reaction Speed Low profile
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.