Learn More
Electron Microscopy Sciences Abh-Jba/Aluminum Chucks
SDP
Supplier:  Electron Microscopy Sciences 7017420
Encompass_Preferred
Machined from solid Aluminum. They come with set screws and a hex key. ABH-JBA Chuck with a three point recessed block holder attachment: for LKB Histo Range and Reichert Super-Cut. Stem measures 11 x 11 mm.
Catalog No. 50-249-75