Learn More
Electron Microscopy Sciences Abh-Lkb/Aluminum Chucks
SDP
Supplier:  Electron Microscopy Sciences 7017414
Encompass_Preferred
Machined from solid Aluminum. They come with set screws and a hex key. ABH-LBK Huxley Chuck. Stem measures 11 x 25 mm.
Catalog No. 50-249-76