Learn More
Aviva Systems Biology Corp Abhd12 Antibody - C-terminal region (ARP44942_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44942_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Abhd12. It was validated on Western Blot. . Alternate names: 1500011G07Rik, 6330583M11Rik, AI431047, AW547313, RP23-241M12.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-169