Learn More
Aviva Systems Biology Corp ABHD12 Antibody - middle region (ARP44941_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44941_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABHD12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ABHD12. Alternate names: ABHD12A, BEM46L2, C20orf22, DKFZP434P106, dJ965G21.2, PHARCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54050