Learn More
Aviva Systems Biology Corp ABHD12B Antibody - middle region (ARP53392_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53392_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABHD12B. It was validated on Western Blot. . Alternate names: BEM46L3, C14orf29, MGC129926, MGC129927, c14_5314ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-834