Learn More
Aviva Systems Biology Corp ABHD13 Antibody - N-terminal region (ARP50137_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50137_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABHD13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ABHD13. Alternate names: BEM46L1, C13orf6, FLJ14906, MGC27058, RP11-153I24.2, bA153I24.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60266