Learn More
Aviva Systems Biology Corp ABHD5 Antibody - N-terminal region (ARP42455_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42455_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ABHD5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ABHD5. Alternate names: CDS, CGI58, IECN2, MGC8731, NCIE2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-58231