Learn More
Aviva Systems Biology Corp ABI3BP Antibody - N-terminal region (ARP47157_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47157_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABI3BP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ABI3BP. Alternate names: FLJ41743, FLJ41754, NESHBP, TARSHApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59476