Learn More
TARGETMOL CHEMICALS INC Abietic Acid 100MG
SDP
Catalog No. 502876501
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Abietic Acid (Sylvic acid) is a diterpene extracted from Pimenta racemosavar. grissea with antimicrobial, antiproliferative and anti-obesity activities. It inhibits lipoxygenase activity and may be studied in allergic diseases. Purity 92.6%
Catalog No. 50-287-6501 Supplier TARGETMOL CHEMICALS INC Supplier No. T3090100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Abietic Acid (Sylvic acid) is a diterpene extracted from Pimenta racemosavar. grissea with antimicrobial, antiproliferative and anti-obesity activities. It inhibits lipoxygenase activity and may be studied in allergic diseases. Purity 92.6%