Learn More
Selleck Chemical LLC Abietic Acid S5122-25mg
SDP
Catalog No. 503112340
Encompass_Preferred
Abietic acid (Sylvic acid Abietate Rosin Acid) an abietane diterpenoid inhibited soybean 5-lipoxygenase with an IC50 of 29 5 1 29 M
Catalog No. 50-311-2340 Supplier Selleck Chemical LLC Supplier No. S512225MG
Abietic acid (Sylvic acid Abietate Rosin Acid) an abietane diterpenoid inhibited soybean 5-lipoxygenase with an IC50 of 29 5 1 29 M