Learn More
Medchemexpress LLC Abietic acid | 514-10-3 | 10 MG
SDP
Catalog No. 5000421010
Encompass_Preferred
Abietic acid, a diterpene from Colophony, demonstrates anti-proliferative, anti-inflammatory, anti-obesity, and bacteriostatic activities. It inhibits lipoxygenase, enhances cell migration, and induces angiogenic potential. It also attenuates sepsis-induced lung injury, aids liver injury recovery, and accelerates wound healing. Research indicates its potential for NSCLC, liver disease, and psoriasis.
  • Diterpene from Colophony.
  • Anti-proliferative and anti-inflammatory properties.
  • Inhibits lipoxygenase activity.
  • Enhances cell migration and angiogenesis.
  • Attenuates sepsis-induced lung injury.
  • Accelerates wound healing.
  • Promising for NSCLC, liver disease, and psoriasis research.
Catalog No. 50-004-21010 Supplier Medchemexpress LLC Supplier No. HYN687110MG
Abietic acid, a diterpene from Colophony, demonstrates anti-proliferative, anti-inflammatory, anti-obesity, and bacteriostatic activities. It inhibits lipoxygenase, enhances cell migration, and induces angiogenic potential. It also attenuates sepsis-induced lung injury, aids liver injury recovery, and accelerates wound healing. Research indicates its potential for NSCLC, liver disease, and psoriasis.
  • Diterpene from Colophony.
  • Anti-proliferative and anti-inflammatory properties.
  • Inhibits lipoxygenase activity.
  • Enhances cell migration and angiogenesis.
  • Attenuates sepsis-induced lung injury.
  • Accelerates wound healing.
  • Promising for NSCLC, liver disease, and psoriasis research.