Learn More
Medchemexpress LLC Abietic acid | 514-10-3 | 25 MG
SDP
Catalog No. 5000421590
Encompass_Preferred
Abietic acid is an orally active diterpene isolated from Colophony. It exhibits anti-proliferative, anti-inflammatory, anti-obesity, bacteriostatic, cell cycle arresting, and pro-apoptotic activities. It is promising for research in non-small-cell lung cancer (NSCLC), liver injury-related diseases, and psoriasis.
  • Inhibits lipoxygenase activity for allergy.
  • Enhances cell migration and tube formation in HUVECs.
  • Induces angiogenic potential through upregulation of extracellular signal-regulated kinase (ERK) and p38 expression.
  • Attenuates sepsis-induced lung injury by inhibiting the NF-κB pathway to inhibit M1 macrophage polarization.
  • Exhibits a positive effect against liver injury by attenuating inflammation and ferroptosis.
  • Significantly reduces the proliferation and growth of NSCLC cells by IKKβ inhibition.
  • Ameliorates psoriasis-like inflammation and modulates gut microbiota in mice.
Catalog No. 50-004-21590 Supplier Medchemexpress LLC Supplier No. 25MGHYN6871
Abietic acid is an orally active diterpene isolated from Colophony. It exhibits anti-proliferative, anti-inflammatory, anti-obesity, bacteriostatic, cell cycle arresting, and pro-apoptotic activities. It is promising for research in non-small-cell lung cancer (NSCLC), liver injury-related diseases, and psoriasis.
  • Inhibits lipoxygenase activity for allergy.
  • Enhances cell migration and tube formation in HUVECs.
  • Induces angiogenic potential through upregulation of extracellular signal-regulated kinase (ERK) and p38 expression.
  • Attenuates sepsis-induced lung injury by inhibiting the NF-κB pathway to inhibit M1 macrophage polarization.
  • Exhibits a positive effect against liver injury by attenuating inflammation and ferroptosis.
  • Significantly reduces the proliferation and growth of NSCLC cells by IKKβ inhibition.
  • Ameliorates psoriasis-like inflammation and modulates gut microbiota in mice.