Learn More
Medchemexpress LLC Abietic acid | 514-10-3 | MFCD00078239 | 50 MG
SDP
Catalog No. 5000419498
Encompass_Preferred
Abietic acid, also known as Abietinic acid or Sylvic acid, is a major component of pine resin.
  • An irritant and contact allergen.
  • Inhibits CYP2B6 with an IC50 of 0.6 μM.
Catalog No. 50-004-19498 Supplier Medchemexpress LLC Supplier No. HYN687150MG
Abietic acid, also known as Abietinic acid or Sylvic acid, is a major component of pine resin.
  • An irritant and contact allergen.
  • Inhibits CYP2B6 with an IC50 of 0.6 μM.