Learn More
Cayman Chemical AbIrateron 10mg
SDP
Catalog No. 502894658
Encompass_Preferred
A CYP17A1 inhibitor (IC50s = 0.0029 and 0.004 µM for the 17,20-lyase and 17α-hydroxylase activities, respectively) and an active metabolite of abiraterone acetate; selective for CYP17A1 over aromatase and 5α-reductase (IC50s = >20 and >50 µM, respectively)
Catalog No. 50-289-4658 Supplier Cayman Chemical Supplier No. 900276810 MG
A CYP17A1 inhibitor (IC50s = 0.0029 and 0.004 µM for the 17,20-lyase and 17α-hydroxylase activities, respectively) and an active metabolite of abiraterone acetate; selective for CYP17A1 over aromatase and 5α-reductase (IC50s = >20 and >50 µM, respectively)