Learn More
Medchemexpress LLC HY-70013 1g , Abiraterone CAS:154229-19-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY700131G
Encompass_Preferred
HY-70013 1g Abiraterone CAS: 154229-19-3 Abiraterone is a potent and irreversible CYP17A1 inhibitor with antiandrogen activity, which inhibits both the 17α-hydroxylase and 17,20-lyase activity of the cytochrome p450 enzyme CYP17 with IC50s of 2.5 nM and 15 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1092