Learn More
TARGETMOL CHEMICALS INC Abiraterone 200MG
SDP
Catalog No. 502879804
Encompass_Preferred
Also available in 1 g, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Abiraterone (CB-7598) (CB-7598) is an effective steroidal cytochrome P450 17alpha-hydroxylase-17, 20-lyase (CYP17) inhibitor (IC50 4 nM). Purity 99.87%
Catalog No. 50-287-9804 Supplier TARGETMOL CHEMICALS INC Supplier No. T6216200MG
Also available in 1 g, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Abiraterone (CB-7598) (CB-7598) is an effective steroidal cytochrome P450 17alpha-hydroxylase-17, 20-lyase (CYP17) inhibitor (IC50 4 nM). Purity 99.87%