Learn More
Medchemexpress LLC Abiraterone acetate | 154229-18-2 | 99.7% | C26H33NO2 | 1 ML
SDP
Catalog No. 5000433647
Encompass_Preferred
Abiraterone acetate (CB7630) is an oral, potent, selective, and irreversible inhibitor of CYP17A1 with antiandrogen activity. It is a proagent form of Abiraterone.
  • Oral, potent, selective, and irreversible inhibitor of CYP17A1
  • Exhibits antiandrogen activity
  • Proagent form of Abiraterone
  • Targets Cytochrome P450
  • Acts on the Metabolic Enzyme/Protease pathway
  • Inhibits human 17,20-lyase and 17α-hydroxylase
Catalog No. 50-004-33647 Supplier Medchemexpress LLC Supplier No. HY7505410MM/1ML
Abiraterone acetate (CB7630) is an oral, potent, selective, and irreversible inhibitor of CYP17A1 with antiandrogen activity. It is a proagent form of Abiraterone.
  • Oral, potent, selective, and irreversible inhibitor of CYP17A1
  • Exhibits antiandrogen activity
  • Proagent form of Abiraterone
  • Targets Cytochrome P450
  • Acts on the Metabolic Enzyme/Protease pathway
  • Inhibits human 17,20-lyase and 17α-hydroxylase