Learn More
Medchemexpress LLC Abiraterone acetate | 154229-18-2 | 99.91% | C26H33NO2 | 5 MG
SDP
Catalog No. 5000433711
Encompass_Preferred
Abiraterone acetate is an oral, potent, selective, and irreversible inhibitor of CYP17A1 with antiandrogen activity. It is a proagent form of Abiraterone.
  • Oral administration
  • Potent, selective, and irreversible inhibitor of CYP17A1
  • Exhibits antiandrogen activity
  • Prodrug form of Abiraterone
  • Inhibits 17,20-lyase and 17α-hydroxylase activity
  • Inhibits proliferation of AR-positive prostate cancer cell lines
  • Blocks synthesis of 5α-dione and DHT
  • Inhibits CRPC progression
  • Prolongs survival in castration-resistant prostate cancer (CRPC)
  • Inhibits dehydroepiandrosterone (DHEA) depletion and Δ4-androstenedione (AD) accumulation
Catalog No. 50-004-33711 Supplier Medchemexpress LLC Supplier No. HY750545MG
Abiraterone acetate is an oral, potent, selective, and irreversible inhibitor of CYP17A1 with antiandrogen activity. It is a proagent form of Abiraterone.
  • Oral administration
  • Potent, selective, and irreversible inhibitor of CYP17A1
  • Exhibits antiandrogen activity
  • Prodrug form of Abiraterone
  • Inhibits 17,20-lyase and 17α-hydroxylase activity
  • Inhibits proliferation of AR-positive prostate cancer cell lines
  • Blocks synthesis of 5α-dione and DHT
  • Inhibits CRPC progression
  • Prolongs survival in castration-resistant prostate cancer (CRPC)
  • Inhibits dehydroepiandrosterone (DHEA) depletion and Δ4-androstenedione (AD) accumulation