Learn More
Medchemexpress LLC Abiraterone acetate | 154229-18-2 | MFCD00934213 | 99.9% | 10 MG
SDP
Catalog No. 5000298805
Encompass_Preferred
Abiraterone acetate analytical standard is the acetate ester prodrug of abiraterone, used as a reference compound in research and analytical assays targeting CYP17A1 activity. It is provided as a highly pure material suitable for identity confirmation, method development, and quantitative analysis.
  • intended for research and analytical applications.
  • validated identity with CAS 154229-18-2.
  • molecular formula C26H33NO2 and molecular weight 391.55.
  • high purity suitable for use as a reference standard.
  • compatible with chromatography and assay development workflows.
Catalog No. 50-002-98805 Supplier Medchemexpress LLC Supplier No. HY75054R10MG
Abiraterone acetate analytical standard is the acetate ester prodrug of abiraterone, used as a reference compound in research and analytical assays targeting CYP17A1 activity. It is provided as a highly pure material suitable for identity confirmation, method development, and quantitative analysis.
  • intended for research and analytical applications.
  • validated identity with CAS 154229-18-2.
  • molecular formula C26H33NO2 and molecular weight 391.55.
  • high purity suitable for use as a reference standard.
  • compatible with chromatography and assay development workflows.