Learn More
Selleck Chemical LLC Abiraterone Acetate S2246-10mM/1mL
SDP
Catalog No. 503111770
Encompass_Preferred
Abiraterone Acetate is an acetate salt form of Abiraterone which is a steroidal cytochrome CYP17 inhibitor with IC50 of 72 nM in a cell-free assay Abiraterone acetate is an oral androgen biosynthesis inhibitor
Catalog No. 50-311-1770 Supplier Selleck Chemical LLC Supplier No. S224610MM/1ML
Abiraterone Acetate is an acetate salt form of Abiraterone which is a steroidal cytochrome CYP17 inhibitor with IC50 of 72 nM in a cell-free assay Abiraterone acetate is an oral androgen biosynthesis inhibitor