Learn More
TARGETMOL CHEMICALS INC ABIRATERONE ACETATE 500MG
SDP
Catalog No. 502579484
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Abiraterone Acetate (CB7630) is an androstene derivative that inhibits STEROID 17-ALPHA-HYDROXYLASE and is used as an ANTINEOPLASTIC AGENT in the treatment of metastatic castration-resistant PROSTATE CANCER. purity: 99%
Catalog No. 50-257-9484 Supplier TARGETMOL CHEMICALS INC Supplier No. T6215500MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Abiraterone Acetate (CB7630) is an androstene derivative that inhibits STEROID 17-ALPHA-HYDROXYLASE and is used as an ANTINEOPLASTIC AGENT in the treatment of metastatic castration-resistant PROSTATE CANCER. purity: 99%