Learn More
Medchemexpress LLC HY-75054 50mg Medchemexpress, Abiraterone acetate CAS:154229-18-2 Purity:>98%
SDP
Catalog No. 502029364
Encompass_Preferred
Medchemexpress, HY-75054 50mg Abiraterone acetate CAS:154229-18-2 Abiraterone acetate (CB7630) is an oral, potent, selective, and irreversible inhibitor of CYP17A1 with antiandrogen activity. Abiraterone acetate is a prodrug form of Abiraterone (CB7598). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9364 Supplier Medchemexpress LLC Supplier No. HY7505450MG
Medchemexpress, HY-75054 50mg Abiraterone acetate CAS:154229-18-2 Abiraterone acetate (CB7630) is an oral, potent, selective, and irreversible inhibitor of CYP17A1 with antiandrogen activity. Abiraterone acetate is a prodrug form of Abiraterone (CB7598). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.