Learn More
Selleck Chemical LLC Abiraterone Acetate 5mg 154229-18-2 CB7630
SDP
Supplier:  Selleck Chemical LLC S22465MG
Encompass_Preferred
Abiraterone Acetate (CB7630) is an acetate salt form of Abiraterone which is a steroidal cytochrome CYP17 inhibitor with IC50 of 72 nM in a cell-free assay. Abiraterone acetate is an oral androgen biosynthesis inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-488-4