Learn More
Medchemexpress LLC Abiraterone-d4 | 2122245-62-7 | 98.0% | C24H27D4NO | 5 MG
SDP
Catalog No. 5000433626
Encompass_Preferred
Abiraterone-d4 is a deuterium-labeled form of Abiraterone, a potent and irreversible CYP17A1 inhibitor with antiandrogen activity. It inhibits both the 17α-hydroxylase and 17,20-lyase activity of the cytochrome p450 enzyme CYP17. This compound can be used as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-33626 Supplier Medchemexpress LLC Supplier No. HY70013S5MG
Abiraterone-d4 is a deuterium-labeled form of Abiraterone, a potent and irreversible CYP17A1 inhibitor with antiandrogen activity. It inhibits both the 17α-hydroxylase and 17,20-lyase activity of the cytochrome p450 enzyme CYP17. This compound can be used as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS