Learn More
Medchemexpress LLC Abiraterone decanoate | 2486052-18-8 | 99.7% | C34H49NO2 | 100 MG
SDP
Catalog No. 5000353326
Encompass_Preferred
Abiraterone decanoate is a potent Abiraterone proagent. It provides a controlled release of Abiraterone and long-acting CYP17 inhibition with intramuscular (IM) delivery.
  • Potent Abiraterone proagent
  • Controlled release of Abiraterone
  • Long-acting CYP17 inhibition
  • Administered via intramuscular (IM) delivery
Catalog No. 50-003-53326 Supplier Medchemexpress LLC Supplier No. HY145786100MG
Abiraterone decanoate is a potent Abiraterone proagent. It provides a controlled release of Abiraterone and long-acting CYP17 inhibition with intramuscular (IM) delivery.
  • Potent Abiraterone proagent
  • Controlled release of Abiraterone
  • Long-acting CYP17 inhibition
  • Administered via intramuscular (IM) delivery