Learn More
Medchemexpress LLC Abiraterone decanoate | 2486052-18-8 | 99.7% | C34H49NO2 | 10MG
SDP
Catalog No. 5000258597
Encompass_Preferred
Abiraterone decanoate is the decanoate ester prodrug of abiraterone designed to provide controlled release and prolonged CYP17 inhibition when formulated for intramuscular delivery. Supplied as a high-purity solid for research and formulation studies; consult the datasheet for purity, storage, and handling details.
  • Provides controlled release of abiraterone for long-acting CYP17 inhibition.
  • High purity suitable for research use (reported 99.69%).
  • Solid (powder) physical form for formulation and preclinical studies.
  • Documented storage: powder stable at -20°C for extended storage; solvent stability conditions provided in datasheet.
  • Intended for use in preclinical and formulation research investigating long-acting androgen biosynthesis inhibition.
Catalog No. 50-002-58597 Supplier Medchemexpress LLC Supplier No. HY14578610MG
Abiraterone decanoate is the decanoate ester prodrug of abiraterone designed to provide controlled release and prolonged CYP17 inhibition when formulated for intramuscular delivery. Supplied as a high-purity solid for research and formulation studies; consult the datasheet for purity, storage, and handling details.
  • Provides controlled release of abiraterone for long-acting CYP17 inhibition.
  • High purity suitable for research use (reported 99.69%).
  • Solid (powder) physical form for formulation and preclinical studies.
  • Documented storage: powder stable at -20°C for extended storage; solvent stability conditions provided in datasheet.
  • Intended for use in preclinical and formulation research investigating long-acting androgen biosynthesis inhibition.