Learn More
Medchemexpress LLC Abiraterone metabolite 1 | 1940176-03-3 | MFCD31619249 | 99.6% | 351.52 g/mol | C24H33NO | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10368710MM 1ML
Encompass_Preferred
Abiraterone metabolite 1 is the 5α-reduced metabolite of abiraterone and a reported CYP17A1 inhibitor. It is supplied as a research reference standard for biochemical, pharmacological, and metabolism studies related to androgen biosynthesis and prostate cancer research.
  • High purity (≈99.6%).
  • Molecular formula C24H33NO and molecular weight 351.52 g/mol.
  • Provided as a 10 mM solution in DMSO (1 ML) and as solid sizes for analytical use.
  • Intended for use as an analytical standard and in vitro biochemical assays.
  • Datasheet, certificate of analysis, and SDS available for quality and safety information.
Catalog No. 50-002-44699