Learn More
Medchemexpress LLC Abiraterone sulfate | 1993430-25-3 | 98.9% | 429.57 | 5 MG
SDP
Catalog No. 5000361848
Encompass_Preferred
Abiraterone sulfate is a metabolite of Abiraterone, a compound recognized for its potent and irreversible CYP17A1 inhibitory and antiandrogen activities. This product is supplied as a solid and is soluble in DMSO, making it suitable for various research applications.
  • Metabolite of Abiraterone
  • Potent and irreversible CYP17A1 inhibitor
  • Exhibits antiandrogen activity
  • Purity of 98.87%
  • Stable under recommended storage conditions
Catalog No. 50-003-61848 Supplier Medchemexpress LLC Supplier No. HY1483755MG
Abiraterone sulfate is a metabolite of Abiraterone, a compound recognized for its potent and irreversible CYP17A1 inhibitory and antiandrogen activities. This product is supplied as a solid and is soluble in DMSO, making it suitable for various research applications.
  • Metabolite of Abiraterone
  • Potent and irreversible CYP17A1 inhibitor
  • Exhibits antiandrogen activity
  • Purity of 98.87%
  • Stable under recommended storage conditions