Learn More
Medchemexpress LLC Abiraterone sulfate | 1993430-25-3 | 98.9% | 429.57 g/mol | C24H31NO4S | 10 MG
SDP
Catalog No. 5000286937
Encompass_Preferred
Abiraterone sulfate is the sulfate metabolite of abiraterone supplied for research use. It is used in studies of drug metabolism and androgen biosynthesis because abiraterone is an irreversible CYP17A1 inhibitor with antiandrogen activity. The compound is provided in small research pack sizes for analytical and pharmacological experiments.
  • Metabolite suitable for drug metabolism studies.
  • Applicable to CYP17A1 and antiandrogen research.
  • High purity for analytical applications (≈98.9%).
  • Chemical formula C24H31NO4S and molecular weight 429.57 g/mol.
  • Available in small research pack sizes, including 10 mg.
Catalog No. 50-002-86937 Supplier Medchemexpress LLC Supplier No. HY14837510MG
Abiraterone sulfate is the sulfate metabolite of abiraterone supplied for research use. It is used in studies of drug metabolism and androgen biosynthesis because abiraterone is an irreversible CYP17A1 inhibitor with antiandrogen activity. The compound is provided in small research pack sizes for analytical and pharmacological experiments.
  • Metabolite suitable for drug metabolism studies.
  • Applicable to CYP17A1 and antiandrogen research.
  • High purity for analytical applications (≈98.9%).
  • Chemical formula C24H31NO4S and molecular weight 429.57 g/mol.
  • Available in small research pack sizes, including 10 mg.