Learn More
Eurofins Discoverx Abl, active, 10 µg. (14-529)
Supplier:  Eurofins Discoverx 14529
Encompass
N-terminal 6xHis-tagged recombinant, human Abl, amino acids 27-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8927