Learn More
Enzo Life Sciences Abl kinase substrate (1mg)
SDP
Catalog No. 502011291
Encompass_Preferred
This peptide, EAIYAAPFAKKK, was identified as a high affinity (Km=4 µM) and specific substrate for the Abl kinase through a degenerate peptide library. The three lysines on the carboxy-terminus allow this peptide to be used in phosphocellulose kinase assays. Purity: ≥95%. Long Term Storage: -20°C.
Catalog No. 50-201-1291 Supplier Enzo Life Sciences Supplier No. BMLP2160001
This peptide, EAIYAAPFAKKK, was identified as a high affinity (Km=4 µM) and specific substrate for the Abl kinase through a degenerate peptide library. The three lysines on the carboxy-terminus allow this peptide to be used in phosphocellulose kinase assays. Purity: ≥95%. Long Term Storage: -20°C.